Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Netherlands
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Bromo-4-chloro-6-methylquinoline
3-Bromo-4-chloro-6-methylquinoline
3-Bromo-4-chloro-6-methylquinoline
3-Bromo-4-chloro-6-methylquinoline
APOSOR309329-1G
:
Apollo Scientific
:
3-Bromo-4-chloro-6-methylquinoline
APOSOR309329-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
Environmentally Preferable:
:
:
256.526 g/mol
:
Ambient
:
1204810-41-2
:
C₁₀H₇BrClN
:
MFCD13192941