Order Entry
Netherlands
ContactUsLinkComponent
1-Oxa-7-azaspiro[3.5]nonane-7-carboxylic acid tert-butyl ester 95+%
1-Oxa-7-azaspiro[3.5]nonane-7-carboxylic acid tert-butyl ester 95+%
Catalog # APOSOR309130-1G
Supplier:  Apollo Scientific
CAS Number:  
1-Oxa-7-azaspiro[3.5]nonane-7-carboxylic acid tert-butyl ester 95+%
Catalog # APOSOR309130-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :227.3 g/mol
Storage Temperature:Ambient
CAS Number:864684-96-8
Formula:C₁₂H₂₁NO₃
MDL Number:MFCD08706105