Methyl (2R)-morpholine-2-carboxylate hydrochloride
Catalog # APOSOR308171-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:181.617 g/mol
Storage Temperature:Ambient
CAS Number:1352709-55-7
Formula:C₆H₁₁NO₃·ClH
MDL Number:MFCD21606371