2-Chloro-3-(3-chloropropyl)-6-methoxyquinoline
Catalog # APOSOR307900-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:270.154 g/mol
Storage Temperature:Ambient
CAS Number:159383-57-0
Formula:C₁₃H₁₃Cl₂NO
MDL Number:MFCD09787635