1-Propyl-1H-pyrazole-5-methanol
Catalog # APOSOR307807-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:140.183 g/mol
Storage Temperature:Ambient
CAS Number:1007517-79-4
Formula:C₇H₁₂N₂O
MDL Number:MFCD08701097