4-(4-Aminophenethyl)piperazine-1-carboxylic acid tert-butyl ester
Catalog # APOSOR307780-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:305.415 g/mol
Storage Temperature:Ambient
CAS Number:329004-08-2
Formula:C₁₇H₂₇N₃O₂
MDL Number:MFCD11590790