3-Amino-3-[1-(tert-butoxycarbonyl)piperidin-4-yl]propanoic acid hydrochloride
Catalog # APOSOR307687-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:308.802 g/mol
Storage Temperature:Ambient
CAS Number:1431365-41-1
Formula:C₁₃H₂₄N₂O₄·ClH
MDL Number:MFCD24368278