Order Entry
Netherlands
ContactUsLinkComponent
diexo-3-Amino-7-oxa-bicyclo[2.2.1]heptane-2-carboxylic acid ethyl ester hydrochloride
diexo-3-Amino-7-oxa-bicyclo[2.2.1]heptane-2-carboxylic acid ethyl ester hydrochloride
Catalog # APOSOR307661-1G
Supplier:  Apollo Scientific
CAS Number:  
diexo-3-Amino-7-oxa-bicyclo[2.2.1]heptane-2-carboxylic acid ethyl ester hydrochloride
Catalog # APOSOR307661-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :221.681 g/mol
Storage Temperature:Ambient
CAS Number:1212145-02-2
Formula:C₉H₁₅NO₃·ClH
MDL Number:MFCD11870780