diexo-3-Amino-7-oxa-bicyclo[2.2.1]hept-5-ene-2-carboxylic acid amide
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:885096-06-0
:
Formula:C₇H₁₀N₂O₂
MDL Number:MFCD09475433
:154.167 g/mol
Storage Temperature:Ambient
Frequently Bought Together