Order Entry
Netherlands
ContactUsLinkComponent
3-Amino-3-benzo[1,3]dioxol-5-yl-propan-1-ol hydrochloride
3-Amino-3-benzo[1,3]dioxol-5-yl-propan-1-ol hydrochloride
Catalog # APOSOR307625-1G
Supplier:  Apollo Scientific
CAS Number:  
3-Amino-3-benzo[1,3]dioxol-5-yl-propan-1-ol hydrochloride
Catalog # APOSOR307625-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :231.676 g/mol
Storage Temperature:Ambient
CAS Number:113511-49-2
Formula:C₁₀H₁₃NO₃·ClH
MDL Number:MFCD06211985