Order Entry
Netherlands
ContactUsLinkComponent
(4-Methoxy-3-methylpyridin-2-yl)methanol 97%
(4-Methoxy-3-methylpyridin-2-yl)methanol 97%
Catalog # APOSOR307212-250MG
Supplier:  Apollo Scientific
CAS Number:  
(4-Methoxy-3-methylpyridin-2-yl)methanol 97%
Catalog # APOSOR307212-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :153.178 g/mol
Storage Temperature:Ambient
CAS Number:86604-77-5
Formula:C₈H₁₁NO₂
MDL Number:MFCD08272123