Order Entry
Netherlands
ContactUsLinkComponent
5-Methyl-1-(propan-2-yl)-1H-pyrazole-3-carboxylic acid
Irritant
5-Methyl-1-(propan-2-yl)-1H-pyrazole-3-carboxylic acid
Catalog # APOSOR306338-250MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
5-Methyl-1-(propan-2-yl)-1H-pyrazole-3-carboxylic acid
Catalog # APOSOR306338-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :168.193 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:50920-49-5
Formula:C₈H₁₂N₂O₂
MDL Number:MFCD08700675