Order Entry
Netherlands
ContactUsLinkComponent
Methyl N2-(tert-butoxycarbonyl)-D-asparaginate
Irritant
Methyl N2-(tert-butoxycarbonyl)-D-asparaginate
Catalog # APOSOR305315-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
Methyl N2-(tert-butoxycarbonyl)-D-asparaginate
Catalog # APOSOR305315-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :246.26 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:147091-87-0
Formula:C₁₀H₁₈N₂O₅
MDL Number:MFCD25541765