
1-Benzothiophen-6-ylmethanol
Catalog # APOSOR305048-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:164.224 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:6179-28-8
Formula:C₉H₈OS
MDL Number:MFCD18451826