Order Entry
Netherlands
ContactUsLinkComponent
6-Methyl-2-azaspiro[3.3]heptane hydrochloride
6-Methyl-2-azaspiro[3.3]heptane hydrochloride
Catalog # APOSOR304407-1G
Supplier:  Apollo Scientific
CAS Number:  
6-Methyl-2-azaspiro[3.3]heptane hydrochloride
Catalog # APOSOR304407-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :147.646 g/mol
Storage Temperature:Ambient
CAS Number:2089649-42-1
Formula:C₇H₁₃N·ClH
MDL Number:MFCD30829398