cis-2-Aminocyclobutan-1-ol hydrochloride
Catalog # APOSOR304384-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:123.581 g/mol
Storage Temperature:Ambient
CAS Number:2070860-49-8
Formula:C₄H₉NO·ClH
MDL Number:MFCD26127763