tert-Butyl trans-3-aminocyclobutane-1-carboxylate
Catalog # APOSOR304347-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:171.237 g/mol
Storage Temperature:Ambient
CAS Number:1192547-86-6
Formula:C₉H₁₇NO₂
MDL Number:MFCD16661182