Order Entry
Netherlands
ContactUsLinkComponent
 
2-[(2S)-Azetidin-2-yl]propan-2-ol hydrochloride
Catalog # APOSOR304337-1G
Supplier:  Apollo Scientific
CAS Number:  
2-[(2S)-Azetidin-2-yl]propan-2-ol hydrochloride
Catalog # APOSOR304337-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :151.634 g/mol
Storage Temperature:Ambient
CAS Number:935669-11-7
Formula:C₆H₁₃NO·ClH
MDL Number:MFCD31617919