Order Entry
Netherlands
Orders LinkContactUsLinkComponent
(R)-(-)-2-Hydroxyhex-5-ene

(R)-(-)-2-Hydroxyhex-5-ene

Flammable
Supplier:   Apollo Scientific
(R)-(-)-2-Hydroxyhex-5-ene
Supplier:  Apollo Scientific
Certificates
(R)-(-)-2-Hydroxyhex-5-ene

2 Options Available Below

Product Details & Documents
  • CAS Number:17397-29-4
  • Formula:C₆H₁₂O
  • MDL Number:MFCD03427201
  • Molecular Weight:100.159 g/mol
  • Boiling Pt:159 °C
  • Density:0.828 g/cm³ (25 °C)
  • Flash Pt:43 °C
  • Storage Temperature:Ambient
  • UN:1987
  • ADR:3,III
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    250 mg
    Log in for availability
    Log in for availability
    1 * 250 mg
    1 * 250 mg - 71,00 €
    71,00 €
    1 * 250 mg
       71,00 €

  • Bottle
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 167,00 €
    167,00 €
    1 * 1 g
       167,00 €