Order Entry
Netherlands
ContactUsLinkComponent
1-(2-Methyl-2,3-dihydro-1H-indol-1-yl)ethan-1-one
1-(2-Methyl-2,3-dihydro-1H-indol-1-yl)ethan-1-one
Catalog # APOSOR303913-500MG
Supplier:  Apollo Scientific
CAS Number:  
1-(2-Methyl-2,3-dihydro-1H-indol-1-yl)ethan-1-one
Catalog # APOSOR303913-500MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :175.227 g/mol
Melting Pt:58…60 °C
Storage Temperature:Ambient
CAS Number:131880-76-7
Formula:C₁₁H₁₃NO
MDL Number:MFCD00456248