4-(1-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)ethyl)morpholine
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1206594-12-8
:
Formula:C₁₈H₂₈BNO₃
MDL Number:MFCD18383925
:317.231 g/mol
Storage Temperature:Refrigerator
Frequently Bought Together