
1-Isopropyl-1H-pyrazole-5-carboxylic acid
Catalog # APOSOR302444-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:154.167 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:920006-32-2
Formula:C₇H₁₀N₂O₂
MDL Number:MFCD03130043