Order Entry
Netherlands
ContactUsLinkComponent
 
7H-Pyrrolo[2,3-d]pyrimidine-5-carbaldehyde 98%
Catalog # APOSOR301310-250MG
Supplier:  Apollo Scientific
CAS Number:  
7H-Pyrrolo[2,3-d]pyrimidine-5-carbaldehyde 98%
Catalog # APOSOR301310-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :147.134 g/mol
Storage Temperature:Ambient
CAS Number:1060815-89-5
Formula:C₇H₅N₃O
MDL Number:MFCD13189361