Bestelling invoeren
Netherlands
ContactUsLinkComponent
1H-Pyrrolo[2,3-b]pyridin-4-amine hydrochloride 97%
1H-Pyrrolo[2,3-b]pyridin-4-amine hydrochloride 97%
Catalog # APOSOR301265-1G
Supplier:  Apollo Scientific
CAS Number:  
1H-Pyrrolo[2,3-b]pyridin-4-amine hydrochloride 97%
Catalog # APOSOR301265-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Verpakking:
    Bottle
  • VPE:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :169.612 g/mol
Storage Temperature:Kamertemperatuur
CAS Number:1134307-94-0
Formula:C₇H₇N₃·ClH
MDL Number:MFCD08689965