Methyl 6-methoxy-2-pyrazinecarboxylate
Catalog # APOSOR301139-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:168.15 g/mol
Storage Temperature:Ambient
CAS Number:23813-24-3
Formula:C₇H₈N₂O₃
MDL Number:MFCD02258380