
1-(5-Bromo-2-chloropyridin-4-yl)ethan-1-one
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1245915-91-6
:
Formula:C₇H₅BrClNO
MDL Number:MFCD11847348
:234.478 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together