
Ethyl {[5-(2,6-dichloropyridin-4-yl)-4-methyl-4H-1,2,4-triazol-3-yl]thio}acetate
Catalog # APOSOR29906-50MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:50 mg
- Seq:2370
Specifications
About this item
:
:347.22 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:680216-57-3
Formula:C₁₂H₁₂Cl₂N₄O₂S
MDL Number:MFCD00833056