
Ethyl 2-methyl-5-(3-nitrophenyl)-3-furoate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:175276-71-8
:
Formula:C₁₄H₁₃NO₅
MDL Number:MFCD00277691
:275.257 g/mol
:GHS07
Storage Temperature:Kamertemperatuur
Frequently Bought Together