Methyl 3-(ethylthio)-6,6-dimethyl-4-oxo-4,5,6,7-tetrahydrobenzo[c]thiophene-1-carboxylate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:172516-42-6
:
Formula:C₁₄H₁₈O₃S₂
MDL Number:MFCD00085119
:298.421 g/mol
Storage Temperature:Ambient
Frequently Bought Together