Order Entry
Netherlands
ContactUsLinkComponent
3-Aminothieno[3,2-d]pyrimidin-4(3H)-one
Irritant
3-Aminothieno[3,2-d]pyrimidin-4(3H)-one
Catalog # APOSOR24373-250MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
3-Aminothieno[3,2-d]pyrimidin-4(3H)-one
Catalog # APOSOR24373-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :167.188 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:648859-53-4
Formula:C₆H₅N₃OS
MDL Number:MFCD00816913