
1-(4-Chlorophenyl)-5-methyl-1H-pyrazole-4-carboxylic acid
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:187998-35-2
:
Formula:C₁₁H₉ClN₂O₂
MDL Number:MFCD01566260
:236.654 g/mol
Melting Pt:209…211 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together