
1-(4-Isothiocyanatophenyl)-1H-pyrazole
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:352018-96-3
:
Formula:C₁₀H₇N₃S
MDL Number:MFCD02681939
:201.248 g/mol
Melting Pt:97…99 °C
Storage Temperature:Ambient
UN:2811
ADR:6.1,III
:GHS07
Frequently Bought Together