Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Netherlands
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Bromo-5-methyl-1-phenyl-1H-pyrazole
4-Bromo-5-methyl-1-phenyl-1H-pyrazole 97%
4-Bromo-5-methyl-1-phenyl-1H-pyrazole 97%
APOSOR23249-1G
:
Apollo Scientific
:
4-Bromo-5-methyl-1-phenyl-1H-pyrazole 97%
APOSOR23249-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
237.096 g/mol
:
37…40 °C
:
GHS07
:
Ambient
:
50877-44-6
:
C₁₀H₉BrN₂
:
MFCD02681923