

1,2-Dimethyl-1H-imidazole-5-sulphonyl chloride
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:849351-92-4
:
Formula:C₅H₇ClN₂O₂S
MDL Number:MFCD03094627
:194.639 g/mol
Melting Pt:44…47 °C
UN:3261
ADR:8,III
:GHS05
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together