Order Entry
Netherlands
ContactUsLinkComponent
1-tert-Butyl 3-ethyl 3-phenylpiperidine-1,3-dicarboxylate fumarate 90%
Irritant
1-tert-Butyl 3-ethyl 3-phenylpiperidine-1,3-dicarboxylate fumarate 90%
Catalog # APOSOR200018-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
1-tert-Butyl 3-ethyl 3-phenylpiperidine-1,3-dicarboxylate fumarate 90%
Catalog # APOSOR200018-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :449.494 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:1980007-98-4
 :1417574-76-5
Formula:C₁₉H₂₇NO₄·C₄H₄O₄
MDL Number:MFCD21333141