Order Entry
Netherlands
ContactUsLinkComponent
3-(4-Isobutylphenyl)-1H-pyrazole
3-(4-Isobutylphenyl)-1H-pyrazole
Catalog # APOSOR183466-1G
Supplier:  Apollo Scientific
CAS Number:  
3-(4-Isobutylphenyl)-1H-pyrazole
Catalog # APOSOR183466-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :200.28 g/mol
Storage Temperature:Ambient
CAS Number:948293-30-9
Formula:C₁₃H₁₆N₂
MDL Number:MFCD08446305