Order Entry
Netherlands
ContactUsLinkComponent
3-(1H-Pyrrol-2-yl)-1H-pyrazole
3-(1H-Pyrrol-2-yl)-1H-pyrazole
Catalog # APOSOR183437-1G
Supplier:  Apollo Scientific
CAS Number:  
3-(1H-Pyrrol-2-yl)-1H-pyrazole
Catalog # APOSOR183437-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :133.151 g/mol
Storage Temperature:Ambient
CAS Number:1019010-39-9
Formula:C₇H₇N₃
MDL Number:MFCD06808816