Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Netherlands
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-(5-Methyl-2-thienyl)-1H-pyrazole
3-(5-Methyl-2-thienyl)-1H-pyrazole
3-(5-Methyl-2-thienyl)-1H-pyrazole
APOSOR183436-1G
:
Apollo Scientific
:
3-(5-Methyl-2-thienyl)-1H-pyrazole
APOSOR183436-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
164.228 g/mol
:
Ambient
:
166334-61-8
:
C₈H₈N₂S
:
MFCD06808810