
4-Methyl-2-thio-1,3-thiazole 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:5685-06-3
Formula:C₄H₅NS₂
MDL Number:MFCD00798571
:131.219 g/mol
Melting Pt:86…89 °C
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together