Order Entry
Netherlands
ContactUsLinkComponent
Methyl 3-bromo-5-iodobenzoate 97%
Irritant
Methyl 3-bromo-5-iodobenzoate 97%
Catalog # APOSOR17113-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
Methyl 3-bromo-5-iodobenzoate 97%
Catalog # APOSOR17113-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :340.941 g/mol
Melting Pt:59…63 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:188813-07-2
Formula:C₈H₆BrIO₂
MDL Number:MFCD00673016