Order Entry
Netherlands
ContactUsLinkComponent
3,7-Dihydro-2-thio-4H-pyrrolo[2,3-d]pyrimidin-4-one
Irritant
3,7-Dihydro-2-thio-4H-pyrrolo[2,3-d]pyrimidin-4-one
Catalog # APOSOR17057-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
3,7-Dihydro-2-thio-4H-pyrrolo[2,3-d]pyrimidin-4-one
Catalog # APOSOR17057-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :167.188 g/mol
 :GHS07
Storage Temperature:Refrigerator
CAS Number:67831-84-9
Formula:C₆H₅N₃OS
MDL Number:MFCD12196502