Order Entry
Netherlands
ContactUsLinkComponent
5-Methyl-1-phenyl-1H-pyrazole-4-boronic acid, pinacol ester 97%
Irritant
5-Methyl-1-phenyl-1H-pyrazole-4-boronic acid, pinacol ester 97%
Catalog # APOSOR1395-250MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
5-Methyl-1-phenyl-1H-pyrazole-4-boronic acid, pinacol ester 97%
Catalog # APOSOR1395-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :284.161 g/mol
Melting Pt:110…112 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:849776-88-1
Formula:C₁₆H₂₁BN₂O₂
MDL Number:MFCD06797463