Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Netherlands
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Amino-1-(2-methylphenyl)ethan-1-ol oxalate
2-Amino-1-(2-methylphenyl)ethan-1-ol oxalate
2-Amino-1-(2-methylphenyl)ethan-1-ol oxalate
APOSOR12965-250MG
:
Apollo Scientific
:
2-Amino-1-(2-methylphenyl)ethan-1-ol oxalate
APOSOR12965-250MG
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
250 mg
Seq:
2590
:
:
241.241 g/mol
:
GHS07
:
Ambient
:
1170238-97-7
:
C₉H₁₃NO·C₂H₂O₄
:
MFCD04117772