
3-[(Pyridin-3-yl)methyl]piperidine-3-carboxylic acid, N-BOC protected
Catalog # APOSOR12918-100MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:320.383 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:887344-18-5
Formula:C₁₇H₂₄N₂O₄
MDL Number:MFCD03840045