

Biphenyl-3-sulphonyl chloride
Catalog # APOSOR12582-100MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:252.717 g/mol
Melting Pt:46…47 °C
Flash Pt:>110 °C
Storage Temperature:Ambient
UN:3261
ADR:8,III
:GHS05
:GHS07
CAS Number:65685-01-0
Formula:C₁₂H₉ClO₂S
MDL Number:MFCD01631880