
3-(4-Bromophenyl)-1-methyl-1H-pyrazole 98%
Catalog # APOSOR12276-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:237.096 g/mol
Melting Pt:79.5…81 °C
:GHS07
Storage Temperature:Ambient
CAS Number:73387-51-6
Formula:C₁₀H₉BrN₂
MDL Number:MFCD01566446