
2-Chloro-6-(1H-pyrazol-1-yl)pyrazine 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:642459-09-4
:
Formula:C₇H₅ClN₄
MDL Number:MFCD09800958
:180.594 g/mol
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together