5-(Hydroxymethyl)-4-(4-methoxyphenyl)-2-(propan-2-yl)-2,4-dihydro-3H-1,2,4-triazol-3-one
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:2244086-24-4
:
Formula:C₁₃H₁₇N₃O₃
MDL Number:MFCD31629932
:263.292 g/mol
Storage Temperature:Ambient
Frequently Bought Together