Ethyl imidazo[1,2-a]pyridine-2-carboxylate hydrobromide
Catalog # APOSOR111305-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:271.111 g/mol
Storage Temperature:Ambient
CAS Number:2549-17-9
Formula:C₁₀H₁₀N₂O₂·BrH
MDL Number:MFCD12545991