Order Entry
Netherlands
ContactUsLinkComponent
1-(4-Chlorophenyl)-5-phenyl-4,5-dihydro-1H-pyrazole-3-carbohydrazide
Corrosive
Irritant
1-(4-Chlorophenyl)-5-phenyl-4,5-dihydro-1H-pyrazole-3-carbohydrazide
Catalog # APOSOR111091-1G
Supplier:  Apollo Scientific
CAS Number:  
Corrosive
Irritant
1-(4-Chlorophenyl)-5-phenyl-4,5-dihydro-1H-pyrazole-3-carbohydrazide
Catalog # APOSOR111091-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :314.77 g/mol
 :GHS05
 :GHS07
Storage Temperature:Ambient
CAS Number:1987070-55-2
Formula:C₁₆H₁₅ClN₄O
MDL Number:MFCD28347988